missing translation for 'onlineSavingsMsg'
Learn More

5-Thio-D-glucose, 97+%, Thermo Scientific Chemicals

Catalog No. p-7042748
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63621MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63621MB Supplier Thermo Scientific Chemicals Supplier No. J63621MB
Only null left

5-Thio-D-glucose is used as an inhibitor of the cellular transport of D-glucose and D-glucose mediated insulin release, it is also an Inhibitor of spermatogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Thio-D-glucose is used as an inhibitor of the cellular transport of D-glucose and D-glucose mediated insulin release, it is also an Inhibitor of spermatogenesis.

Solubility
Miscible with water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 20408-97-3
Molecular Formula C6H12O5S
Molecular Weight (g/mol) 196.217
MDL Number MFCD00006663
InChI Key IJJLRUSZMLMXCN-SLPGGIOYSA-N
Synonym 5-thio-d-glucose, d-glucose, 5-thio, alpha-d-glucothiopyranose, 2r,3r,4s,5r-2,3,4,6-tetrahydroxy-5-mercaptohexanal, unii-3prv1384uo, 5-thio-alpha-d-glucopyranose, glucose, 5-thio-, d, 2r,3r,4s,5r-2,3,4,6-tetrahydroxy-5-sulfanylhexanal
PubChem CID 88527
IUPAC Name (2R,3R,4S,5R)-2,3,4,6-tetrahydroxy-5-sulfanylhexanal
SMILES C(C(C(C(C(C=O)O)O)O)S)O

Specifications

Melting Point 135°C to 138°C
Color White
Quantity 25 mg
Beilstein 1865193
Merck Index 14,9334
Solubility Information Miscible with water.
Formula Weight 196.22
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 5-Thio-D-glucose
EINECSNumber 243-798-8
RTECSNumber LZ7500000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only