missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butylisophthalic Acid 99.0+%, TCI America™

Catalog No. B138025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B138025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B138025G Supplier TCI America Supplier No. B138025G
Only null left

Chemical Identifiers

CAS 2359-09-3
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00067047
InChI Key BJLUCDZIWWSFIB-UHFFFAOYSA-N
PubChem CID 75379
IUPAC Name 5-tert-butylbenzene-1,3-dicarboxylic acid
SMILES CC(C)(C)C1=CC(=CC(=C1)C(=O)O)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 222.24
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-tert-Butylisophthalic Acid