missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate Hydrate 98.0+%, TCI America™

Catalog No. B31675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31675G 5 g
B316725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31675G Supplier TCI America Supplier No. B31675G

Chemical Identifiers

CAS 71989-18-9
Molecular Formula C24H27NO6
Molecular Weight (g/mol) 425.481
MDL Number MFCD00037135
InChI Key OTKXCALUHMPIGM-FQEVSTJZSA-N
Synonym fmoc-glu otbu-oh, fmoc-l-glutamic acid 5-tert-butyl ester, 2s-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, n-fmoc-l-glutamic acid 5-tert-butyl ester, fmoc-glu obut, fmoc-glu tbu, fmoc-l-glu otbu, pubchem12950, fmoc-l-glu otbu-oh
PubChem CID 2724637
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 90°C
Color White
Quantity 5 g
Formula Weight 425.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate Hydrate