missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamate Hydrate 98.0+%, TCI America™
$39.14 - $111.84
Chemical Identifiers
| CAS | 104091-08-9 |
|---|---|
| Molecular Formula | C24H27NO6 |
| Molecular Weight (g/mol) | 425.48 |
| MDL Number | MFCD00077055 |
| InChI Key | OTKXCALUHMPIGM-UHFFFAOYNA-N |
| Synonym | fmoc-d-glu otbu-oh, fmoc-d-glutamic acid 5-tert-butyl ester, fmoc-d-glu obut-oh, fmoc-d-glutamic acid gamma-tert-butyl ester, commercial, 2r-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, ambotzfaa1324, pubchem10015, fmoc-d-glu tbu-oh, fmoc-d-glu otbu-oh h |
| PubChem CID | 7018822 |
| IUPAC Name | 5-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoic acid |
| SMILES | CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Chemical Identifiers
| 104091-08-9 | |
| 425.48 | |
| OTKXCALUHMPIGM-UHFFFAOYNA-N | |
| 7018822 | |
| CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C24H27NO6 | |
| MFCD00077055 | |
| fmoc-d-glu otbu-oh, fmoc-d-glutamic acid 5-tert-butyl ester, fmoc-d-glu obut-oh, fmoc-d-glutamic acid gamma-tert-butyl ester, commercial, 2r-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, ambotzfaa1324, pubchem10015, fmoc-d-glu tbu-oh, fmoc-d-glu otbu-oh h | |
| 5-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoic acid |
Specifications
| 104091-08-9 | |
| White | |
| MFCD00077055 | |
| fmoc-d-glu otbu-oh, fmoc-d-glutamic acid 5-tert-butyl ester, fmoc-d-glu obut-oh, fmoc-d-glutamic acid gamma-tert-butyl ester, commercial, 2r-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, ambotzfaa1324, pubchem10015, fmoc-d-glu tbu-oh, fmoc-d-glu otbu-oh h | |
| CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 425.48 | |
| 425.48 | |
| Crystalline Powder |
| 92°C | |
| C24H27NO6 | |
| 1 g | |
| OTKXCALUHMPIGM-UHFFFAOYNA-N | |
| 5-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoic acid | |
| 7018822 | |
| ≥98.0% (HPLC,T) | |
| 5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamate Hydrate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator