missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamate Hydrate 98.0+%, TCI America™

Catalog No. B36691G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36691G 1 g
B36695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36691G Supplier TCI America Supplier No. B36691G
Only null left

Chemical Identifiers

CAS 104091-08-9
Molecular Formula C24H27NO6
Molecular Weight (g/mol) 425.48
MDL Number MFCD00077055
InChI Key OTKXCALUHMPIGM-UHFFFAOYNA-N
Synonym fmoc-d-glu otbu-oh, fmoc-d-glutamic acid 5-tert-butyl ester, fmoc-d-glu obut-oh, fmoc-d-glutamic acid gamma-tert-butyl ester, commercial, 2r-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, ambotzfaa1324, pubchem10015, fmoc-d-glu tbu-oh, fmoc-d-glu otbu-oh h
PubChem CID 7018822
IUPAC Name 5-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 92°C
Color White
Quantity 1 g
Formula Weight 425.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamate Hydrate
TSCA No
Recommended Storage Refrigerator