missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butyl L-Glutamate Hydrate 98.0+%, TCI America™

Catalog No. B40451G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40451G 1 g
B40455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40451G Supplier TCI America Supplier No. B40451G

Chemical Identifiers

CAS 2419-56-9
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00038580
InChI Key OIOAKXPMBIZAHL-LURJTMIESA-N
PubChem CID 7010573
IUPAC Name (2S)-2-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)N

Specifications

Melting Point 182°C
Color White
Quantity 1 g
Synonym h-glu otbu-oh, l-glutamic acid 5-tert-butyl ester, h-glu o-t-bu-oh, 2s-2-amino-5-tert-butoxy-5-oxopentanoic acid, l-gamma-t-butyl glutamate, l-glutamic acid,5-1,1-dimethylethyl ester, 5-tert-butyl l-glutamate, l-glutamic acid, 5-1,1-dimethylethyl ester, glu otbu, glu tbu
Formula Weight 203.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-tert-Butyl L-Glutamate Hydrate
TSCA No
Recommended Storage Refrigerator