missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butyl-2-methyl-3-furoic acid, 97%

Catalog No. aaa1925802
Change view
Click to view available options
Quantity:
0.5 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1925802 0.5 g
AAA1925888 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1925802 Supplier Thermo Scientific Chemicals Supplier No. A1925802
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38422-62-7 | C10H14O3 | 182.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38422-62-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00221068
InChI Key NWWAPYOVUAEIJB-UHFFFAOYSA-N
Synonym 5-tert-butyl-2-methylfuran-3-carboxylic acid, 5-tert-butyl-2-methyl-3-furoic acid, maybridge3_006912, acmc-20am1c, 5-tert-butyl-3-carboxy-2-methylfuran, 5-? 1,?1-?dimethylethyl ?-?2-?methyl-3-?furancarboxylic acid
PubChem CID 2742176
IUPAC Name 5-tert-butyl-2-methylfuran-3-carboxylic acid
SMILES CC1=C(C=C(O1)C(C)(C)C)C(=O)O

Specifications

Melting Point 90°C to 94°C
Quantity 0.5 g
Formula Weight 182.22
Percent Purity 97%
Chemical Name or Material 5-tert-Butyl-2-methyl-3-furoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only