missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butyl 1-Methyl L-Glutamate Hydrochloride 98.0+%, TCI America™

Catalog No. p-7123091
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36701G 1 g
B36705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36701G Supplier TCI America Supplier No. B36701G

Chemical Identifiers

CAS 6234-01-1
Molecular Formula C10H20ClNO4
Molecular Weight (g/mol) 253.72
MDL Number MFCD00153438
InChI Key YIFPACFSZQWAQF-RZUXQYDTNA-N
Synonym h-glu otbu-ome.hcl, l-glutamic acid 5-tert-butyl 1-methyl ester hydrochloride, h-glu otbu-ome hcl, 5-tert-butyl 1-methyl l-glutamate hydrochloride, h-glu otbu-ome inverted exclamation mark currencyhcl, h-glu otbu-ome hydrochloride, s-5-tert-butyl 1-methyl 2-aminopentanedioate hydrochloride, h-glu otbu-omehcl, 5-tert-butyl 1-methyl 2s-2-aminopentanedioate hydrochloride, glu otbu-ome.hcl
PubChem CID 16218751
IUPAC Name 5-O-tert-butyl 1-O-methyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CC(C)(C)OC(=O)CCC(C(=O)OC)N.Cl

Specifications

Melting Point 128°C
Color White
Quantity 1 g
Formula Weight 253.72
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-tert-Butyl 1-Methyl L-Glutamate Hydrochloride
TSCA No
Recommended Storage Refrigerator