missing translation for 'onlineSavingsMsg'
Learn More

5-(tert-Butoxycarbonylamino)-1-pentanol 97.0+%, TCI America™

Catalog No. B28695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28695G 5 g
B286925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28695G Supplier TCI America Supplier No. B28695G

Chemical Identifiers

CAS 75178-90-4
Molecular Formula C10H21NO3
Molecular Weight (g/mol) 203.28
MDL Number MFCD01862955
InChI Key DDGNGFVNTZJMMZ-UHFFFAOYSA-N
Synonym 5-boc-amino-1-pentanol, tert-butyl n-5-hydroxypentyl carbamate, tert-butyl 5-hydroxypentyl carbamate, 5-tert-butoxycarbonylamino-1-pentanol, n-5-hydroxypentyl carbamic acid tert-butyl ester, tert-butyl 5-hydroxypentylcarbamate, carbamic acid, 5-hydroxypentyl-, 1,1-dimethylethyl ester, n-boc-5-aminopentanol, 5-boc-amino 1-pentanol, acmc-209ox9
PubChem CID 4456136
IUPAC Name tert-butyl N-(5-hydroxypentyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCO

Specifications

Color Colorless
Quantity 5 g
Formula Weight 203.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-(tert-Butoxycarbonylamino)-1-pentanol