missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfo-4'-diethylamino-2,2'-dihydroxyazobenzene [Reagent for Aluminum], TCI America™

Catalog No. S01281G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S01281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01281G Supplier TCI America Supplier No. S01281G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1563-01-5
Molecular Formula C16H19N3O5S
Molecular Weight (g/mol) 365.404
MDL Number MFCD00024960
InChI Key DGIJTEMQAXADQJ-GHRIWEEISA-N
Synonym DDB, 4′C-Diethylamino-2,2′C-dihydroxy-5-sulfoazobenzene
PubChem CID 6115900
IUPAC Name 3-[(2E)-2-[4-(diethylamino)-6-oxocyclohexa-2,4-dien-1-ylidene]hydrazinyl]-4-hydroxybenzenesulfonic acid
SMILES CCN(CC)C1=CC(=O)C(=NNC2=C(C=CC(=C2)S(=O)(=O)O)O)C=C1

Specifications

Color Purple-Red
Quantity 1 g
Formula Weight 365.40
Physical Form Crystalline Powder
Chemical Name or Material 5-Sulfo-4′-diethylamino-2,2′-dihydroxyazobenzene [Reagent for Aluminum]
RTECSNumber DB6050000
TSCA Yes