missing translation for 'onlineSavingsMsg'
Learn More

5-Quinoxalinol 98.0+%, TCI America™

$42.36 - $150.16

Chemical Identifiers

CAS 17056-99-4
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD00972004
InChI Key YDXPVVWMFFHSPT-UHFFFAOYSA-N
Synonym quinoxalin-5-ol, 5-hydroxyquinoxaline, 5-quinoxalinol, quinoxaline-5-ol, quinoxalin-5 1h-one, ksc172c0d, zuqddqfxsnxeod-uhfffaoysa-n
PubChem CID 12522983
IUPAC Name 1H-quinoxalin-5-one
SMILES C1=CC(=O)C2=NC=CNC2=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
Q0095200MG
View Documents
TCI America
Q0095200MG
200 mg
Each for $42.36
Only null left
 
Q00951G
View Documents
TCI America
Q00951G
1 g
Each for $150.16
Only null left
 
Description

Description

Specifications

Chemical Identifiers

17056-99-4
146.149
YDXPVVWMFFHSPT-UHFFFAOYSA-N
12522983
C1=CC(=O)C2=NC=CNC2=C1
C8H6N2O
MFCD00972004
quinoxalin-5-ol, 5-hydroxyquinoxaline, 5-quinoxalinol, quinoxaline-5-ol, quinoxalin-5 1h-one, ksc172c0d, zuqddqfxsnxeod-uhfffaoysa-n
1H-quinoxalin-5-one

Specifications

17056-99-4
Yellow
C8H6N2O
200 mg
YDXPVVWMFFHSPT-UHFFFAOYSA-N
1H-quinoxalin-5-one
12522983
≥98.0% (GC,T)
5-Quinoxalinol
101°C
184°C
MFCD00972004
quinoxalin-5-ol, 5-hydroxyquinoxaline, 5-quinoxalinol, quinoxaline-5-ol, quinoxalin-5 1h-one, ksc172c0d, zuqddqfxsnxeod-uhfffaoysa-n
C1=CC(=O)C2=NC=CNC2=C1
146.149
146.15
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents