missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Quinolinecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America Q00891G
Specifications
| 5-Quinolinecarboxylic Acid | |
| 342°C | |
| MFCD00047608 | |
| 5-quinolinecarboxylic acid, quinoline-5-carboxylicacid, quinoline-5-carboxylicacid,98+%, zlchem 983, pubchem19952, 5-quinolinecarboxylicacid, acmc-209on5, ksc173e0j, 5-quinolinecarboxylic acid 6ci,7ci,8ci,9ci, quinoline-5-carboxylicacidethylester;5-quinoline-carboxylic acid ethyl ester | |
| OC(=O)C1=C2C=CC=NC2=CC=C1 | |
| 173.17 | |
| 173.17 | |
| Crystalline Powder |
| 7250-53-5 | |
| C10H7NO2 | |
| 1 g | |
| RAYMXZBXQCGRGX-UHFFFAOYSA-N | |
| quinoline-5-carboxylic acid | |
| 232489 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 7250-53-5 | |
| 173.17 | |
| RAYMXZBXQCGRGX-UHFFFAOYSA-N | |
| 232489 | |
| OC(=O)C1=C2C=CC=NC2=CC=C1 |
| C10H7NO2 | |
| MFCD00047608 | |
| 5-quinolinecarboxylic acid, quinoline-5-carboxylicacid, quinoline-5-carboxylicacid,98+%, zlchem 983, pubchem19952, 5-quinolinecarboxylicacid, acmc-209on5, ksc173e0j, 5-quinolinecarboxylic acid 6ci,7ci,8ci,9ci, quinoline-5-carboxylicacidethylester;5-quinoline-carboxylic acid ethyl ester | |
| quinoline-5-carboxylic acid |
Safety and Handling
TSCA : No