missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrid-4-ylthiophene-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-3629714
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC67001CB 250mg Amber Glass Bottle
CC67001DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC67001CB Supplier Thermo Scientific Chemicals Supplier No. CC67001CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 216867-32-2
Molecular Formula C10H7NO2S
Molecular Weight (g/mol) 205.23
MDL Number MFCD07376007
InChI Key OJOWGMLSJKRJNV-UHFFFAOYSA-N
Synonym 5-pyridin-4-yl thiophene-2-carboxylic acid, 4-2-carboxythien-5-yl pyridine, 5-pyrid-4-ylthiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 5-4-pyridinyl, 5-4-pyridyl-2-thiophenecarboxylic acid, 5-4-pyridyl thiophene-2-carboxylic acid, 5-4-pyridinyl-2-thiophenecarboxylic acid, 5-pyridin-4-yl-thiophene-2-carboxylic acid, 5-pyridin-4-yl thiophene-2-carboxylic aci, 2-thiophenecarboxylic acid, 5-4-pyridinyl
PubChem CID 21928576
IUPAC Name 5-pyridin-4-ylthiophene-2-carboxylic acid
SMILES OC(=O)C1=CC=C(S1)C1=CC=NC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 205.24
Percent Purity 97%
Chemical Name or Material 5-Pyrid-4-ylthiophene-2-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.