missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrid-3-ylthiophene-2-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC66801DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC66801DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

278803-20-6
205.231
HENDYXUZFUWIMG-UHFFFAOYSA-N
4739100
C1=CC(=CN=C1)C2=CC=C(S2)C(=O)O
C10H7NO2S
MFCD06496471
5-pyrid-3-ylthiophene-2-carboxylic acid, 5-pyridin-3-yl thiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 5-3-pyridinyl, 5-3-pyridyl thiophene-2-carboxylic acid, 5-pyridin-3-yl-thiophene-2-carboxylic acid, 5-3-pyridinyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-3-pyridinyl
5-pyridin-3-ylthiophene-2-carboxylic acid

Specifications

5-Pyrid-3-ylthiophene-2-carboxylic acid
96.5
97%
C10H7NO2S
MFCD06496471
HENDYXUZFUWIMG-UHFFFAOYSA-N
5-pyridin-3-ylthiophene-2-carboxylic acid
4739100
97%
278803-20-6
100.0
Amber Glass Bottle
1g
5-pyrid-3-ylthiophene-2-carboxylic acid, 5-pyridin-3-yl thiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 5-3-pyridinyl, 5-3-pyridyl thiophene-2-carboxylic acid, 5-pyridin-3-yl-thiophene-2-carboxylic acid, 5-3-pyridinyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-3-pyridinyl
C1=CC(=CN=C1)C2=CC=C(S2)C(=O)O
205.231
205.24
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions