missing translation for 'onlineSavingsMsg'
Learn More

5-Piperidinothiophene-2-carbaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 24372-48-3
Molecular Formula C10H13NOS
Molecular Weight (g/mol) 195.28
MDL Number MFCD05263177
InChI Key SFDXRMCPPLZJBG-UHFFFAOYSA-N
Synonym 5-piperidin-1-yl thiophene-2-carbaldehyde, 5-piperidin-1-yl-thiophene-2-carbaldehyde, 5-piperidino-2-thiophenecarbaldehyde, 5-piperidin-1-yl-thiophene-2-carboxaldehyde, 5-piperidylthiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-1-piperidinyl, piperidinothiophenecarbaldehyde, 5-piperidin-1-yl-2-formylthiophene, 2-thiophenecarboxaldehyde,5-1-piperidinyl
PubChem CID 2113341
IUPAC Name 5-piperidin-1-ylthiophene-2-carbaldehyde
SMILES O=CC1=CC=C(S1)N1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC72604CB
SDS View Documents
Thermo Scientific Chemicals
CC72604CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC72604DA
SDS View Documents
Thermo Scientific Chemicals
CC72604DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

24372-48-3
195.28
SFDXRMCPPLZJBG-UHFFFAOYSA-N
2113341
O=CC1=CC=C(S1)N1CCCCC1
C10H13NOS
MFCD05263177
5-piperidin-1-yl thiophene-2-carbaldehyde, 5-piperidin-1-yl-thiophene-2-carbaldehyde, 5-piperidino-2-thiophenecarbaldehyde, 5-piperidin-1-yl-thiophene-2-carboxaldehyde, 5-piperidylthiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-1-piperidinyl, piperidinothiophenecarbaldehyde, 5-piperidin-1-yl-2-formylthiophene, 2-thiophenecarboxaldehyde,5-1-piperidinyl
5-piperidin-1-ylthiophene-2-carbaldehyde

Specifications

24372-48-3
100.0
Amber Glass Bottle
MFCD05263177
5-piperidin-1-yl thiophene-2-carbaldehyde, 5-piperidin-1-yl-thiophene-2-carbaldehyde, 5-piperidino-2-thiophenecarbaldehyde, 5-piperidin-1-yl-thiophene-2-carboxaldehyde, 5-piperidylthiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-1-piperidinyl, piperidinothiophenecarbaldehyde, 5-piperidin-1-yl-2-formylthiophene, 2-thiophenecarboxaldehyde,5-1-piperidinyl
O=CC1=CC=C(S1)N1CCCCC1
195.28
195.28
5-Piperidinothiophene-2-carbaldehyde
96.5
97%
C10H13NOS
250mg
SFDXRMCPPLZJBG-UHFFFAOYSA-N
5-piperidin-1-ylthiophene-2-carbaldehyde
2113341
97%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'