missing translation for 'onlineSavingsMsg'
Learn More

5-Phenylthiophene-2-carboxylic acid, 95+%, Thermo Scientific™

Catalog No. CC10501DE
Change view
Click to view available options
Quantity:
250mg
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC10501DE 5g Amber Glass Bottle
CC10501CB 250mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC10501DE Supplier Thermo Scientific Chemicals Supplier No. CC10501DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19163-24-7
Molecular Formula C11H8O2S
Molecular Weight (g/mol) 204.243
InChI Key QGMFBCDNJUZQBZ-UHFFFAOYSA-N
Synonym 5-phenyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-phenyl, 5-phenyl-2-thiophene carboxylic acid, pubchem19533, 5-phenyl-2-thenoic acid, 2-carboxy-5-phenylthiophene, 5-phenyl 2-thiophenecarboxylic acid, 5-phenyl-thiophene-2-carboxylic acid
PubChem CID 736494
IUPAC Name 5-phenylthiophene-2-carboxylic acid
SMILES C1=CC=C(C=C1)C2=CC=C(S2)C(=O)O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5g
Formula Weight 204.25
Percent Purity 95%
Chemical Name or Material 5-Phenylthiophene-2-carboxylic acid