missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyltetrazole, 99%

Catalog No. AC263720050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC263720050 5 g Glass bottle
AC263720250 25 g Glass bottle
AC263721000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC263720050 Supplier Thermo Scientific Chemicals Supplier No. 263720050

CAS: 18039-42-4 | C7H6N4 | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18039-42-4
Molecular Formula C7H6N4
Molecular Weight (g/mol) 146.15
MDL Number MFCD00022388
InChI Key MARUHZGHZWCEQU-UHFFFAOYSA-N
Synonym 5-phenyl-1h-tetrazole, 5-phenyltetrazole, 1h-tetrazole, 5-phenyl, 2h-tetrazole, 5-phenyl, phenyltetrazole, 5-phenyl tetrazole, expandex 5pt, expandex ox 5pt, kempore 50xpt, 5-phenyltetrazole van
PubChem CID 87425
IUPAC Name 5-phenyl-2H-tetrazole
SMILES C1=CC=C(C=C1)C2=NNN=N2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 146.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenyltetrazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable solid.
Harmful if swallowed.
Risk of explosion if heated under confinement.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-950-8

RUO – Research Use Only