missing translation for 'onlineSavingsMsg'
Learn More

5-Phenylpyridine-2-carboxylic acid, 95%, Thermo Scientific™

Catalog No. CC55001DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC55001DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC55001DA Supplier Thermo Scientific Chemicals Supplier No. CC55001DA

Chemical Identifiers

CAS 75754-04-0
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD04114189
InChI Key CNCJXSKAKGXNKJ-UHFFFAOYSA-N
Synonym 5-phenylpicolinic acid, 2-pyridinecarboxylic acid, 5-phenyl, 5-phenylpicolinicacid, pubchem22107, 5-phenyl-picolinic acid, acmc-1bb5a, 2-carboxy-5-phenylpyridine, 3-phenyl-6-carboxypyridine, 5-phenyl-2-pyridinecarboxylic acid, 5-phenyl-pyridine-2-carboxylic acid
PubChem CID 2762870
IUPAC Name 5-phenylpyridine-2-carboxylic acid
SMILES C1=CC=C(C=C1)C2=CN=C(C=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 5-Phenylpyridine-2-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.