missing translation for 'onlineSavingsMsg'
Learn More

5-Phenylisoxazole 98.0+%, TCI America™

Catalog No. P23091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23091G 1 g
P23095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23091G Supplier TCI America Supplier No. P23091G
Only null left

Chemical Identifiers

CAS 1006-67-3
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00090038
InChI Key BXQDLEHCXQQSCH-UHFFFAOYSA-N
PubChem CID 136800
IUPAC Name 5-phenyl-1,2-oxazole
SMILES C1=CC=C(C=C1)C2=CC=NO2

Specifications

Melting Point 22°C
Color White
Boiling Point 260°C
Quantity 1 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Phenylisoxazole