missing translation for 'onlineSavingsMsg'
Learn More

5-Phenylcyclohexane-1,3-dione, 97%

Catalog No. AAA1213606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1213606 5 g
AAA1213614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1213606 Supplier Thermo Scientific Chemicals Supplier No. A1213606
Only null left

CAS: 493-72-1 | C12H12O2 | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 493-72-1
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.23
MDL Number MFCD00051846
InChI Key ZFVDSCASCRHXCP-SNVBAGLBSA-N
PubChem CID 568962
IUPAC Name 5-phenylcyclohexane-1,3-dione
SMILES OC1=CC(=O)C[C@@H](C1)C1=CC=CC=C1

Specifications

Melting Point ∼185°C (decomposition)
Quantity 5 g
Beilstein 1103470
Synonym 5-phenyl-1,3-cyclohexanedione, 5-phenyl-cyclohexane-1,3-dione, 1,3-cyclohexanedione, 5-phenyl, acmc-1anml, 5-phenylcyclohexan-1,3-dione, 1,3-dioxo-5-phenylcyclohexane, 5phenylcyclohexane-1, 3-dione, 1,3-cyclohexanedione,5-phenyl, 5-phenyl-cyclohexane-1, 3-dione
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material 5-Phenylcyclohexane-1,3-dione
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only