missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Phenyl-3-(trifluoromethyl)-1H-pyrazole 98.0+%, TCI America™
$78.93 - $242.57
Chemical Identifiers
| CAS | 4027-54-7 |
|---|---|
| Molecular Formula | C10H7F3N2 |
| Molecular Weight (g/mol) | 212.18 |
| MDL Number | MFCD00156064 |
| InChI Key | KJKWJOAARUHGNU-UHFFFAOYSA-N |
| PubChem CID | 611322 |
| IUPAC Name | 3-phenyl-5-(trifluoromethyl)-1H-pyrazole |
| SMILES | FC(F)(F)C1=CC(=NN1)C1=CC=CC=C1 |
Chemical Identifiers
| 4027-54-7 | |
| 212.18 | |
| KJKWJOAARUHGNU-UHFFFAOYSA-N | |
| 3-phenyl-5-(trifluoromethyl)-1H-pyrazole |
| C10H7F3N2 | |
| MFCD00156064 | |
| 611322 | |
| FC(F)(F)C1=CC(=NN1)C1=CC=CC=C1 |
Specifications
| 4027-54-7 | |
| White-Yellow | |
| MFCD00156064 | |
| KJKWJOAARUHGNU-UHFFFAOYSA-N | |
| 3-phenyl-5-(trifluoromethyl)-1H-pyrazole | |
| 611322 | |
| ≥98.0% (GC) | |
| 5-Phenyl-3-(trifluoromethyl)-1H-pyrazole |
| 124°C | |
| C10H7F3N2 | |
| 1 g | |
| FC(F)(F)C1=CC(=NN1)C1=CC=CC=C1 | |
| 212.18 | |
| 212.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No