missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-3-(trifluoromethyl)-1H-pyrazole 98.0+%, TCI America™

Catalog No. P17565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17565G 5 g
P17561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17565G Supplier TCI America Supplier No. P17565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4027-54-7
Molecular Formula C10H7F3N2
Molecular Weight (g/mol) 212.18
MDL Number MFCD00156064
InChI Key KJKWJOAARUHGNU-UHFFFAOYSA-N
PubChem CID 611322
IUPAC Name 3-phenyl-5-(trifluoromethyl)-1H-pyrazole
SMILES FC(F)(F)C1=CC(=NN1)C1=CC=CC=C1

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 5 g
Formula Weight 212.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenyl-3-(trifluoromethyl)-1H-pyrazole