missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Phenyl-3-(trifluoromethyl)-1H-pyrazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 124°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 212.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Phenyl-3-(trifluoromethyl)-1H-pyrazole |
Safety and Handling
| TSCA | No |