missing translation for 'onlineSavingsMsg'
Learn More

(5-Phenyl-1,3-oxazol-4-yl)methanol, ≥95%, Thermo Scientific™

Catalog No. p-2693784
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC10209CB 250mg Amber Glass Bottle
CC10209DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC10209CB Supplier Thermo Scientific Chemicals Supplier No. CC10209CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 352018-88-3
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD03086105
InChI Key LVHWTAMRDRDXJP-UHFFFAOYSA-N
Synonym 5-phenyloxazol-4-yl methanol, 5-phenyl-1,3-oxazol-4-yl methanol, 5-phenyl-1,3-oxazole-4-methanol, 4-oxazolemethanol,5-phenyl, 5-phenyl-4-oxazolyl methanol, 5-phenyl-oxazol-4-yl-methanol, 5-phenyl-1,3-oxazol-4-yl methan-1-ol
PubChem CID 2776302
IUPAC Name (5-phenyl-1,3-oxazol-4-yl)methanol
SMILES C1=CC=C(C=C1)C2=C(N=CO2)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 175.19
Percent Purity 95%
Chemical Name or Material (5-Phenyl-1, 3-oxazol-4-yl)methanol