missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5-Phenyl-1,3-oxazol-4-yl)methanol, ≥95%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 94.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 95% |
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 175.19 |
| Percent Purity | 95% |
| Chemical Name or Material | (5-Phenyl-1, 3-oxazol-4-yl)methanol |