missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5 Percent Rhodium on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst, Spectrum™ Chemical
$408.54 - $8215.96
Chemical Identifiers
| CAS | 7440-16-6 |
|---|---|
| Molecular Formula | Rh |
| Molecular Weight (g/mol) | 102.91 |
| MDL Number | MFCD00011201 MFCD03458392 |
| InChI Key | MHOVAHRLVXNVSD-UHFFFAOYSA-N |
| IUPAC Name | rhodium |
| SMILES | [Rh] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-607-970
|
Spectrum Chemical Mfg Cor
SP3012G |
2 g |
Each for $408.54
|
|
|||||
|
18-607-971
|
Spectrum Chemical Mfg Cor
SP30110G |
10 g |
Each for $1,853.61
|
|
|||||
|
18-607-972
|
Spectrum Chemical Mfg Cor
SP30150G |
50 g |
Each for $8,215.96
|
|
|||||
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.