missing translation for 'onlineSavingsMsg'
Learn More

5 Percent Rhodium on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst, Spectrum™ Chemical

Catalog No. 18607970
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18607970 2 g
18607971 10 g
18607972 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607970 Supplier Spectrum Chemical Mfg Cor Supplier No. SP3012G
Get Up to $500 in Chemicals with Purchase
View all promotions
  • BASF catalyst containing reduced 5% rhodium compound with particle size of 27 microns

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
IUPAC Name rhodium
SMILES [Rh]

Specifications

Physical Form Powder
Quantity 2 g
Assay Percent Range 5%
Concentration or Composition (by Analyte or Components) 5% Rhodium on Alumina

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.