missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5 Percent Rhodium on Activated Carbon Powder, Reduced, Water Wet, 27 microns, BASF Catalyst, Spectrum™ Chemical
$134.43 - $3398.90
Chemical Identifiers
| CAS | 7440-16-6 |
|---|---|
| Molecular Formula | Rh |
| Molecular Weight (g/mol) | 102.91 |
| MDL Number | MFCD00011201 MFCD03458392 |
| InChI Key | MHOVAHRLVXNVSD-UHFFFAOYSA-N |
| IUPAC Name | rhodium |
| SMILES | [Rh] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-607-966
|
Spectrum Chemical Mfg Cor
SP3001G |
1 g |
Each for $134.43
|
|
|||||
|
18-607-967
|
Spectrum Chemical Mfg Cor
SP3005G |
5 g |
Each for $519.73
|
|
|||||
|
18-607-968
|
Spectrum Chemical Mfg Cor
SP30010G |
10 g |
Each for $810.47
|
|
|||||
|
18-607-969
|
Spectrum Chemical Mfg Cor
SP30050G |
50 g |
Each for $3,398.90
|
|
|||||
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.