missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Oxohexanoic Acid 98.0+%, TCI America™
Supplier: TCI America K001425G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C6H10O3 | |
| MFCD00004412 | |
| 4-acetylbutyric acid, 5-ketohexanoic acid, hexanoic acid, 5-oxo, 5-ketocaproic acid, 5-oxo-hexanoic acid, 4-acetylbutyricacid, 5-keto-n-caproic acid, 5-oxocaproic acid, delta-oxocaproic acid, delta-ketocaproic acid | |
| CHEBI:15888 | |
| CC(=O)CCCC(=O)O |
Specifications
| 5-Oxohexanoic Acid | |
| 14°C | |
| C6H10O3 | |
| MFCD00004412 | |
| MGTZCLMLSSAXLD-UHFFFAOYSA-N | |
| 5-oxohexanoic acid | |
| 18407 | |
| 130.14 | |
| Liquid |
| 3128-06-1 | |
| 160°C | |
| 25 g | |
| 4-acetylbutyric acid, 5-ketohexanoic acid, hexanoic acid, 5-oxo, 5-ketocaproic acid, 5-oxo-hexanoic acid, 4-acetylbutyricacid, 5-keto-n-caproic acid, 5-oxocaproic acid, delta-oxocaproic acid, delta-ketocaproic acid | |
| CC(=O)CCCC(=O)O | |
| 130.14 | |
| CHEBI:15888 | |
| ≥98.0% (GC,T) |
Safety and Handling
EINECSNumber : (2)-1495&(2)-1503
TSCA : Yes