missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-N2-isobutyryl-2'-deoxyguanosine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals J6423203
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-N2-isobutyryl-2'-deoxyguanosine | |
| MFCD15145247 | |
| 657.69 |
| C35H36FN5O7 | |
| 1g | |
| 98% |
Chemical Identifiers
| C35H36FN5O7 | |
| MFCD15145247 |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO â Research Use Only