missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7133923
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
N06675G 5 g
N066725G 25 g
2 options

Catalog No. N06675G

Supplier: TCI America N06675G

Only null left

Chemical Identifiers

CAS 120-74-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00085356
InChI Key FYGUSUBEMUKACF-UHFFFAOYSA-N
Synonym 5-norbornene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, norbornenecarboxylic acid, exo-5-norbornene-2-carboxylic acid, 5-norbornene-2-carboxylic acid, exo, 5-norbornene-2-carboxylic acid 8ci, bicyclo 2.2.1 hept-2-ene-5-carboxylic acid, bicyclo 2.2.1-5-heptene-2-carboxylic acid, norborn-5-ene-2-carboxylic acid
PubChem CID 78949
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carboxylic acid
SMILES C1C2CC(C1C=C2)C(=O)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Norbornene-2-carboxylic Acid