missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxylic acid, predominantly endo isomer, 97%

Catalog No. p-7044846
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0844406 5 g
AAL0844414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0844406 Supplier Thermo Scientific Chemicals Supplier No. L0844406

CAS: 120-74-1 | C8H10O2 | 138.166 g/mol

5-Norbornene-2-carboxylic acid is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Norbornene-2-carboxylic acid is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible with water.

Notes
Incompatible with oxidizing agents, bases. Store away from oxidizing agents.

Chemical Identifiers

CAS 120-74-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00085356
InChI Key FYGUSUBEMUKACF-UHFFFAOYSA-N
Synonym 5-norbornene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, norbornenecarboxylic acid, exo-5-norbornene-2-carboxylic acid, 5-norbornene-2-carboxylic acid, exo, 5-norbornene-2-carboxylic acid 8ci, bicyclo 2.2.1 hept-2-ene-5-carboxylic acid, bicyclo 2.2.1-5-heptene-2-carboxylic acid, norborn-5-ene-2-carboxylic acid
PubChem CID 78949
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carboxylic acid
SMILES C1C2CC(C1C=C2)C(=O)O

Specifications

Density 1.129
Boiling Point 136°C to 138°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.494
Quantity 5 g
UN Number UN3265
Beilstein 637167
Solubility Information Not miscible with water.
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 5-Norbornene-2-carboxylic acid, predominantly endo isomer
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 204-422-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only