missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Norbornene-2-carboxylic acid, 98%, mixture of isomers, Thermo Scientific™
Chemical Identifiers
| CAS | 120-74-1 |
|---|---|
| Molecular Formula | C8H10O2 |
| Molecular Weight (g/mol) | 138.16 |
| InChI Key | FYGUSUBEMUKACF-UHFFFAOYSA-N |
| Synonym | 5-norbornene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, norbornenecarboxylic acid, exo-5-norbornene-2-carboxylic acid, 5-norbornene-2-carboxylic acid, exo, 5-norbornene-2-carboxylic acid 8ci, bicyclo 2.2.1 hept-2-ene-5-carboxylic acid, bicyclo 2.2.1-5-heptene-2-carboxylic acid, norborn-5-ene-2-carboxylic acid |
| PubChem CID | 78949 |
| IUPAC Name | bicyclo[2.2.1]hept-2-ene-5-carboxylic acid |
| SMILES | C1C2CC(C1C=C2)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC453301000
|
Thermo Scientific Chemicals
453301000 |
100g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 120-74-1 | |
| 138.16 | |
| 5-norbornene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, norbornenecarboxylic acid, exo-5-norbornene-2-carboxylic acid, 5-norbornene-2-carboxylic acid, exo, 5-norbornene-2-carboxylic acid 8ci, bicyclo 2.2.1 hept-2-ene-5-carboxylic acid, bicyclo 2.2.1-5-heptene-2-carboxylic acid, norborn-5-ene-2-carboxylic acid | |
| bicyclo[2.2.1]hept-2-ene-5-carboxylic acid |
| C8H10O2 | |
| FYGUSUBEMUKACF-UHFFFAOYSA-N | |
| 78949 | |
| C1C2CC(C1C=C2)C(=O)O |
Specifications
| 120-74-1 | |
| 136.0°C to 138.0°C (14.0mmHg) | |
| Authentic | |
| C8H10O2 | |
| 1.129 | |
| FYGUSUBEMUKACF-UHFFFAOYSA-N | |
| bicyclo[2.2.1]hept-2-ene-5-carboxylic acid | |
| 78949 | |
| 98% |
| 1.1290g/mL | |
| >110°C | |
| 97.5% min. (total of isomers) (GC) | |
| 100g | |
| 5-norbornene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, norbornenecarboxylic acid, exo-5-norbornene-2-carboxylic acid, 5-norbornene-2-carboxylic acid, exo, 5-norbornene-2-carboxylic acid 8ci, bicyclo 2.2.1 hept-2-ene-5-carboxylic acid, bicyclo 2.2.1-5-heptene-2-carboxylic acid, norborn-5-ene-2-carboxylic acid | |
| C1C2CC(C1C=C2)C(=O)O | |
| 138.16 | |
| 138.16 | |
| 5-Norbornene-2-carboxylic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 204-422-8
RUO â Research Use Only