missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxylic acid, 98%, mixture of endo and exo

Catalog No. AC453300250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453300250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC453300250 Supplier Thermo Scientific Chemicals Supplier No. 453300250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 120-74-1 | C8H10O2 | 138.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120-74-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.16
InChI Key FYGUSUBEMUKACF-UHFFFAOYSA-N
Synonym 5-norbornene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, norbornenecarboxylic acid, exo-5-norbornene-2-carboxylic acid, 5-norbornene-2-carboxylic acid, exo, 5-norbornene-2-carboxylic acid 8ci, bicyclo 2.2.1 hept-2-ene-5-carboxylic acid, bicyclo 2.2.1-5-heptene-2-carboxylic acid, norborn-5-ene-2-carboxylic acid
PubChem CID 78949
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carboxylic acid
SMILES C1C2CC(C1C=C2)C(=O)O

Specifications

Color Colorless to Yellow
Density 1.1290g/mL
Boiling Point 136.0°C to 138.0°C (14.0 mmHg)
Flash Point >110°C
Assay Percent Range 97.5% min. (total of isomers) (GC)
Odor Stench
Refractive Index 1.4930 to 1.4960 (20°C, 589 nm)
Quantity 25 g
UN Number 3265
Specific Gravity 1.129
Formula Weight 138.16
Percent Purity 98%
Concentration or Composition (by Analyte or Components) Bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Physical Form Liquid or Solid
Chemical Name or Material 5-Norbornene-2-Carboxylic Acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
DOTInformation Hazard Class 8
EINECSNumber 204-422-8

RUO – Research Use Only