missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxaldehyde 95.0+%, TCI America™

Catalog No. p-7133845
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05045ML 5 mL
N050425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05045ML Supplier TCI America Supplier No. N05045ML
Only null left

Chemical Identifiers

CAS 5453-80-5
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00167568
InChI Key AJIBZRIAUXVGQJ-UHFFFAOYSA-N
Synonym 5-norbornene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carbaldehyde, 2-formyl-5-norbornene, 5-formylbicyclohept-2-ene, 5-norbornene-2-carbaldehyde, 5-norbornane-2-carboxaldehyde, bicyclo 2.2.1 hept-2-ene-5-carbaldehyde
PubChem CID 95117
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carbaldehyde
SMILES C1C2CC(C1C=C2)C=O

Specifications

Color Yellow
Boiling Point 72°C
Quantity 5 mL
UN Number 1989
Formula Weight 122.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Norbornene-2-carboxaldehyde