missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxaldehyde, 95%, mixture of endo and exo

CAS: 5453-80-5 | C8H10O | 122.17 g/mol

Supplier:  Thermo Scientific Chemicals 149100250

 View more versions of this product

Catalog No. AC149100250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Norbornene-2-carboxaldehyde
5453-80-5
100.0
67°C to 70°C (12.0 mmHg)
Authentic
Glass bottle
1.4870 to 1.492
25 g
5-norbornene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carbaldehyde, 2-formyl-5-norbornene, 5-formylbicyclohept-2-ene, 5-norbornene-2-carbaldehyde, 5-norbornane-2-carboxaldehyde, bicyclo 2.2.1 hept-2-ene-5-carbaldehyde
AJIBZRIAUXVGQJ-UHFFFAOYSA-N
bicyclo[2.2.1]hept-2-ene-5-carbaldehyde
95117
95%
90-95%
90.0
1.0180g/mL
50.6°C
94% min. (GC)
C8H10O
MFCD00167568
1.018
Solubility in water: insoluble
C1C2CC(C1C=C2)C=O
122.17
122.17
Powder

Chemical Identifiers

5453-80-5
122.17
AJIBZRIAUXVGQJ-UHFFFAOYSA-N
95117
C1C2CC(C1C=C2)C=O
C8H10O
MFCD00167568
5-norbornene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carbaldehyde, 2-formyl-5-norbornene, 5-formylbicyclohept-2-ene, 5-norbornene-2-carbaldehyde, 5-norbornane-2-carboxaldehyde, bicyclo 2.2.1 hept-2-ene-5-carbaldehyde
bicyclo[2.2.1]hept-2-ene-5-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep container tightly closed.

GHS Signal Word: Warning

EINECSNumber : 226-698-9

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only