missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxaldehyde, 95%, mixture of endo and exo

Catalog No. AC149100050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149100050 5 g Glass bottle
AC149100250 25 g Glass bottle
AC149101000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC149100050 Supplier Thermo Scientific Chemicals Supplier No. 149100050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5453-80-5 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5453-80-5
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00167568
InChI Key AJIBZRIAUXVGQJ-UHFFFAOYSA-N
Synonym 5-norbornene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carbaldehyde, 2-formyl-5-norbornene, 5-formylbicyclohept-2-ene, 5-norbornene-2-carbaldehyde, 5-norbornane-2-carboxaldehyde, bicyclo 2.2.1 hept-2-ene-5-carbaldehyde
PubChem CID 95117
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carbaldehyde
SMILES C1C2CC(C1C=C2)C=O

Specifications

CAS Min % 90.0
CAS Max % 100.0
Density 1.0180g/mL
Boiling Point 67°C to 70°C (12.0 mmHg)
Flash Point 50.6°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4870 to 1.492
Quantity 5 g
Specific Gravity 1.018
Solubility Information Solubility in water: insoluble
Formula Weight 122.17
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Norbornene-2-carboxaldehyde, 90-95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep container tightly closed.
EINECSNumber 226-698-9
TSCA TSCA

RUO – Research Use Only