missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carbonitrile (mixture of isomers) 98.0+%, TCI America™

Catalog No. p-7134016
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08975G 5 g
N089725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08975G Supplier TCI America Supplier No. N08975G

Chemical Identifiers

CAS 95-11-4
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.167
MDL Number MFCD00167563
InChI Key BMAXQTDMWYDIJX-UHFFFAOYSA-N
Synonym 5-norbornene-2-carbonitrile, bicyclo 2.2.1 hept-5-ene-2-carbonitrile, 5-norbornene-2-carbonitrile, endo, 5-cyano-2-norbornene, 5-cyanobicyclo 2.2.1 hept-2-ene, 5-norbornene-2-carbonitrile, mixture of isomers, endo-2-cyanobicyclo 2-2-1 hept-5-ene, bicyclo 2.2.1 hept-5-ene-2-carbonitrile, endo, bicyclo 2.2.1 hept-2-ene-5-carbonitrile
PubChem CID 7218
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carbonitrile
SMILES C1C2CC(C1C=C2)C#N

Specifications

Melting Point 10°C
Color Yellow
Quantity 5 g
Formula Weight 119.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Norbornene-2-carbonitrile (mixture of isomers)
EINECSNumber (4)-1214
RTECSNumber RB7930000
TSCA No