missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2,3-dimethanol (mixture of endo- and exo-, predominantly endo-isomer) 95.0+%, TCI America™

Catalog No. N07645G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07645G 5 g
N07641G 1 g
N076425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N07645G Supplier TCI America Supplier No. N07645G

Chemical Identifiers

CAS 85-39-2
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
InChI Key IGHHPVIMEQGKNE-UHFFFAOYSA-N
Synonym Bicyclo[2.2.1]hept-5-ene-2,3-dimethanol, 2,3-Bis(hydroxymethyl)-5-norbornene
PubChem CID 97724
IUPAC Name [3-(hydroxymethyl)-2-bicyclo[2.2.1]hept-5-enyl]methanol
SMILES C1C2C=CC1C(C2CO)CO

Specifications

Melting Point 82°C
Color White
Quantity 5 g
Formula Weight 154.21
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Norbornene-2,3-dimethanol (mixture of endo- and exo-, predominantly endo-isomer)