missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornen-2-ol, mixture of endo and exo, 98+%

CAS: 13080-90-5 | C7H10O | 110.156 g/mol

Supplier:  Thermo Scientific Chemicals H5699406

 View more versions of this product

Catalog No. AAH5699406


Only null left
Explore available promotions

Description

Description

It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).

Solubility
Slightly soluble in water

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
Specifications

Specifications

5-Norbornen-2-ol
13080-90-5
51°C (124°F)
MFCD00167566
UN1325
Slightly soluble in water
C1C2CC(C1C=C2)O
110.156
110.15
mixture of endo and exo
104°C to 106°C
C7H10O
5 g
5-norbornene-2-ol, bicyclo 2.2.1 hept-5-en-2-ol, 5-norbornen-2-ol, 5-norbornen-2-ol, exo, exo-2-norbornenol, bicyclo 2.2.1 hept-2-en-5-ol, bicyclo 2.2.1 hept-5-en-2-ol, exo, 5-norbornen-2-ol exo
MKOSBHNWXFSHSW-UHFFFAOYSA-N
bicyclo[2.2.1]hept-2-en-5-ol
96066
99%

Chemical Identifiers

13080-90-5
110.156
MKOSBHNWXFSHSW-UHFFFAOYSA-N
96066
C1C2CC(C1C=C2)O
C7H10O
MFCD00167566
5-norbornene-2-ol, bicyclo 2.2.1 hept-5-en-2-ol, 5-norbornen-2-ol, 5-norbornen-2-ol, exo, exo-2-norbornenol, bicyclo 2.2.1 hept-2-en-5-ol, bicyclo 2.2.1 hept-5-en-2-ol, exo, 5-norbornen-2-ol exo
bicyclo[2.2.1]hept-2-en-5-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 235-987-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only