missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornen-2-methanol (mixture of isomers) 98.0+%, TCI America™

Catalog No. N09795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09795G 5 g
N097925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09795G Supplier TCI America Supplier No. N09795G
Only null left

Chemical Identifiers

CAS 95-12-5
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00167571
InChI Key LUMNWCHHXDUKFI-UHFFFAOYSA-N
Synonym 5-norbornene-2-methanol, bicyclo 2.2.1 hept-5-en-2-ylmethanol, bicyclo 2.2.1 hept-5-ene-2-methanol, cyclol, 2-hydroxymethyl-5-norbornene, 5-hydroxymethyl-2-norbornene, 5-norbornene 2-methanol, 5-hydroxymethylbicyclo 2.2.1 hept-2-ene, 2-hydroxymethyl bicyclo 2.2.1 hept-5-ene
PubChem CID 78946
IUPAC Name 5-bicyclo[2.2.1]hept-2-enylmethanol
SMILES C1C2CC(C1C=C2)CO

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 124.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Norbornen-2-methanol (mixture of isomers)