missing translation for 'onlineSavingsMsg'
Learn More

5-Nonanone 98.0+%, TCI America™

Catalog No. p-7133737
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
N029425ML 25 mL
N0294100ML 100 mL
N0294500ML 500 mL
3 options

Catalog No. N029425ML

Supplier: TCI America N029425ML

Only null left

Chemical Identifiers

CAS 502-56-7
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00009484
InChI Key WSGCRAOTEDLMFQ-UHFFFAOYSA-N
Synonym 5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
PubChem CID 10405
IUPAC Name nonan-5-one
SMILES CCCCC(=O)CCCC

Specifications

Color Yellow
Boiling Point 186°C
Quantity 25 mL
UN Number 1224
Formula Weight 142.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Nonanone
EINECSNumber (2)-0572&(2)-2475
RTECSNumber RA8230000
TSCA Yes