missing translation for 'onlineSavingsMsg'
Learn More

5-Nonanone, 98%

CAS: 502-56-7 | C9H18O | 142.24 g/mol

$56.03 - $56.03

Chemical Identifiers

CAS 502-56-7
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00009484
InChI Key WSGCRAOTEDLMFQ-UHFFFAOYSA-N
Synonym 5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
PubChem CID 10405
IUPAC Name nonan-5-one
SMILES CCCCC(=O)CCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC159760250
SDS View Documents
Thermo Scientific Chemicals
159760250
25 mL Glass bottle
Each for $56.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

502-56-7
142.24
WSGCRAOTEDLMFQ-UHFFFAOYSA-N
10405
CCCCC(=O)CCCC
C9H18O
MFCD00009484
5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
nonan-5-one

Specifications

502-56-7
100.0
Colorless to Yellow
186.0°C to 187.0°C
Authentic
Glass bottle
1.4180 to 1.4200
MFCD00009484
01, I, 365
5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
WSGCRAOTEDLMFQ-UHFFFAOYSA-N
nonan-5-one
10405
98%
5-Nonanone, 98%
97.5
-50.0°C
0.8200g/mL
65°C
97.5% min. (GC)
C9H18O
CH3(CH2)3CO(CH2)3CH3
25 mL
0.82
Solubility in water: insoluble. Other solubilities: soluble in ethanol;, very soluble in diethyl ether, chloroform.
CCCCC(=O)CCCC
142.24
142.24
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-946-5

RTECSNumber : RA8230000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only