missing translation for 'onlineSavingsMsg'
Learn More

5-Nonanone, 98%

Catalog No. AC159760250
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159760250 25 mL Glass bottle
AC159761000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159760250 Supplier Thermo Scientific Chemicals Supplier No. 15976 0250

CAS: 502-56-7 | C9H18O | 142.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 502-56-7
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00009484
InChI Key WSGCRAOTEDLMFQ-UHFFFAOYSA-N
Synonym 5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
PubChem CID 10405
IUPAC Name nonan-5-one
SMILES CCCCC(=O)CCCC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -50.0°C
Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 186.0°C to 187.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4180 to 1.4200
Linear Formula CH3(CH2)3CO(CH2)3CH3
Quantity 25 mL
Beilstein 01, I, 365
Specific Gravity 0.82
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol;, very soluble in diethyl ether, chloroform.
Formula Weight 142.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Nonanone, 98%
Show More Show Less
EINECSNumber 207-946-5
RTECSNumber RA8230000
TSCA TSCA

RUO – Research Use Only