missing translation for 'onlineSavingsMsg'
Learn More

5-Nonanone, 98%

Catalog No. AAA1372222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1372222 100 g
AAA1372236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1372222 Supplier Thermo Scientific Chemicals Supplier No. A1372222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 502-56-7 | C9H18O | 142.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 502-56-7
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00009484
InChI Key WSGCRAOTEDLMFQ-UHFFFAOYSA-N
Synonym 5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
PubChem CID 10405
IUPAC Name nonan-5-one
SMILES CCCCC(=O)CCCC

Specifications

Melting Point -50°C
Density 0.824
Boiling Point 186°C to 187°C
Flash Point 65°C (149°F)
Refractive Index 1.419
Quantity 100 g
Beilstein 1743583
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material 5-Nonanone
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 207-946-5
RTECSNumber RA8230000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only