missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrouracil 99.0+%, TCI America™

Catalog No. p-7133726
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02815G 5 g
N028125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02815G Supplier TCI America Supplier No. N02815G
Only null left

Chemical Identifiers

CAS 611-08-5
Molecular Formula C4H3N3O4
Molecular Weight (g/mol) 157.09
MDL Number MFCD00006021
InChI Key TUARVSWVPPVUGS-UHFFFAOYSA-N
Synonym 5-nitrouracil, 2,4-dihydroxy-5-nitropyrimidine, 5-nitropyrimidine-2,4-diol, uracil, 5-nitro, 5-nitropyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-nitro, 5-nitro-1,2,3,4-tetrahydropyrimidine-2,4-dione, 4-hydroxy-5-nitropyrimidin-2 1h-one, 2,4 1h,3h-pyrimidinedione-5-nitro, 5-nitrouracil 2,4-dihydroxy-5-nitropyrimidine
PubChem CID 69135
ChEBI CHEBI:60763
IUPAC Name 5-nitro-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES [O-][N+](=O)C1=CNC(=O)NC1=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 157.09
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitrouracil
RTECSNumber UV9104545
TSCA Yes