missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrouracil, 98+%

Catalog No. p-7082154
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1244814 25 g
AAA1244822 100 g
AAA1244836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1244814 Supplier Thermo Scientific Chemicals Supplier No. A1244814
Only null left

CAS: 611-08-5 | C4H3N3O4 | 157.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-08-5
Molecular Formula C4H3N3O4
Molecular Weight (g/mol) 157.09
MDL Number MFCD00006021
InChI Key TUARVSWVPPVUGS-UHFFFAOYSA-N
Synonym 5-nitrouracil, 2,4-dihydroxy-5-nitropyrimidine, 5-nitropyrimidine-2,4-diol, uracil, 5-nitro, 5-nitropyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-nitro, 5-nitro-1,2,3,4-tetrahydropyrimidine-2,4-dione, 4-hydroxy-5-nitropyrimidin-2 1h-one, 2,4 1h,3h-pyrimidinedione-5-nitro, 5-nitrouracil 2,4-dihydroxy-5-nitropyrimidine
PubChem CID 69135
ChEBI CHEBI:60763
IUPAC Name 5-nitro-1H-pyrimidine-2,4-dione
SMILES [O-][N+](=O)C1=CNC(=O)NC1=O

Specifications

Melting Point ∼300°C
Quantity 25 g
Beilstein 10410
Formula Weight 157.09
Percent Purity ≥98%
Chemical Name or Material 5-Nitrouracil
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-250-4
RTECSNumber UV9104545
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only