missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrothiophene-2-carboxaldehyde, 98%

Catalog No. AAL0149306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0149306 5 g
AAL0149314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0149306 Supplier Thermo Scientific Chemicals Supplier No. L0149306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4521-33-9 | C5H3NO3S | 157.143 g/mol

5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 4521-33-9
Molecular Formula C5H3NO3S
Molecular Weight (g/mol) 157.143
MDL Number MFCD00005433
InChI Key CHTSWZNXEKOLPM-UHFFFAOYSA-N
Synonym 5-nitrothiophene-2-carboxaldehyde, 5-nitro-2-thiophenecarboxaldehyde, 5-nitro-2-thiophenecarbaldehyde, 5-nitro-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-nitro, 5-nitrothiophene-2-aldehyde, thiophenecarboxaldehyde, 5-nitro, pubchem15668, 5-nitrothiophen aldehyde, 5-nitro-2-formyl-thiophene
PubChem CID 78281
IUPAC Name 5-nitrothiophene-2-carbaldehyde
SMILES C1=C(SC(=C1)[N+](=O)[O-])C=O

Specifications

Melting Point 72°C to 76°C
Quantity 5 g
Beilstein 120540
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Formula Weight 157.15
Percent Purity 98%
Chemical Name or Material 5-Nitrothiophene-2-carboxaldehyde
EINECSNumber 224-850-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only