missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrothiophene-2-carbonitrile, 98+%

Catalog No. AAL1105806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1105806 5 g
AAL1105803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1105806 Supplier Thermo Scientific Chemicals Supplier No. L1105806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16689-02-4 | C5H2N2O2S | 154.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16689-02-4
Molecular Formula C5H2N2O2S
Molecular Weight (g/mol) 154.143
MDL Number MFCD00052401
InChI Key FLYONFCGDKAMIH-UHFFFAOYSA-N
Synonym 5-nitro-2-thiophenecarbonitrile, 5-cyano-2-nitrothiophene, 2-cyano-5-nitrothiophene, 5-nitrothiophene-2-carbonitirile, 5-nitro-thiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-nitro, zlchem 478, maybridge1_005980, 5-nitro-2-cyanothiophene, acmc-209dv8
PubChem CID 519294
IUPAC Name 5-nitrothiophene-2-carbonitrile
SMILES C1=C(SC(=C1)[N+](=O)[O-])C#N

Specifications

Melting Point 43°C to 45°C
Quantity 5 g
UN Number UN3439
Beilstein 123934
Formula Weight 154.15
Percent Purity ≥98%
Chemical Name or Material 5-Nitrothiophene-2-carbonitrile
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only