missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitroisatin 98.0+%, TCI America™
$71.23 - $277.23
Chemical Identifiers
| CAS | 611-09-6 |
|---|---|
| Molecular Formula | C8H4N2O4 |
| Molecular Weight (g/mol) | 192.13 |
| MDL Number | MFCD00005720 |
| InChI Key | UNMYHYODJHKLOC-UHFFFAOYSA-N |
| Synonym | 5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione |
| PubChem CID | 4669250 |
| IUPAC Name | 5-nitro-1H-indole-2,3-dione |
| SMILES | C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2 |
Chemical Identifiers
| 611-09-6 | |
| 192.13 | |
| UNMYHYODJHKLOC-UHFFFAOYSA-N | |
| 4669250 | |
| C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2 |
| C8H4N2O4 | |
| MFCD00005720 | |
| 5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione | |
| 5-nitro-1H-indole-2,3-dione |
Specifications
| 611-09-6 | |
| C8H4N2O4 | |
| 5 g | |
| UNMYHYODJHKLOC-UHFFFAOYSA-N | |
| 5-nitro-1H-indole-2,3-dione | |
| 4669250 | |
| ≥98.0% (HPLC,T) | |
| 5-Nitroisatin |
| 255°C | |
| MFCD00005720 | |
| 5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione | |
| C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2 | |
| 192.13 | |
| 192.13 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : NL7970000
TSCA : No